missing translation for 'onlineSavingsMsg'
Learn More

Melford EDTA Disodium Salt

Product Code. 30018256
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
5 kg
Unit Size:
1kg
250g
500g
5kg
Product Code. Quantity unitSize
30018256 250 g 250g
30018255 1 kg 1kg
30018258 5 kg 5kg
30018257 500 g 500g
4 options
This item is not returnable. View return policy

Product Code. 30018256

Brand: Melford E57020250.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

CAS Number: 6381-92-6

Molecular Formula: C10H14N2O8Na2 2H2O

Molecular Weight: 372.2

Specifications

CAS 6381-92-6
Molecular Formula C10H14N2O8Na2-2H2O
Quantity 250 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.