missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford EDTA Disodium Salt
Click to view available options
Quantity:
250 g
500 g
1 kg
5 kg
Unit Size:
1kg
250g
500g
5kg
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
CAS Number: 6381-92-6
Molecular Formula: C10H14N2O8Na2 2H2O
Molecular Weight: 372.2
Specifications
Specifications
| CAS | 6381-92-6 |
| Molecular Formula | C10H14N2O8Na2-2H2O |
| Quantity | 1 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction