missing translation for 'onlineSavingsMsg'
Learn More

EDTA dipotassium magnesium salt, For complexometry, ≥98%, Honeywell Fluka™

Product Code. 15693940 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
6 x 100 g
50 kg
Unit Size:
100g
50kg
Pack of 6
This item is not returnable. View return policy

Product Code. 15693940

Brand: Honeywell-Fluka 33350100G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

for complexometry,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15708-48-2
Molecular Formula C10H12K2MgN2O8
Molecular Weight (g/mol) 390.714
MDL Number MFCD00054450
InChI Key MUEOBEUHFKBRJH-UHFFFAOYSA-J
Synonym dipotassium magnesium ethylenediaminetetraacetate, magnesate 2-, n,n'-1,2-ethanediylbis n-carboxy-.kappa.o methyl glycinato-.kappa.n,.kappa.o 4--, dipotassium, oc-6-21, edta dipotassium magnesium salt monohydrate, ethylenediaminetetraacetic acid dipotassium magnesium salt, ethylenediamine-n,n,n',n'-tetraacetic acid dipotassium magnesium salt, edta, dipotassium magnesium salt, magnesate 2-, n,n'-1,2-ethanediylbis n-carboxy-kappao methyl glycinato-kappan,kappao 4--, dipotassium, oc-6-21, magnesate 2-, n,n'-1,2-ethanediylbis n-carboxy-kappao methyl glycinato-kappan,kappao 4--, potassium 1:2 , oc-6-21, magnesium 2+ dipotassium ion ion 4-edta, dipotassium n,n'-ethylenebis n-carboxymethyl glycinato 4--n,n',o,o',on,on' magnesate 2-
PubChem CID 85061
IUPAC Name magnesium;dipotassium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].[Mg+2].[K+].[K+]

Specifications

Quantity 100 g
UN Number NONH for all modes of transport
Formula Weight 426.74g/mol
Percent Purity ≥98%
Chemical Name or Material Ethylenediaminetetraacetic acid dipotassium magnesium salt
Product Identifier
  • EDTA dipotassium magnesium salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.