missing translation for 'onlineSavingsMsg'
Learn More

EDTA Diammonium Salt, ≥99%, Honeywell™ Fluka™

Product Code. 15610690 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15610690 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15610690 Supplier Honeywell-Fluka Supplier No. 03640500G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 20824-56-0
Quantity 500 g
Molecular Formula C10H24N4O9
Linear Formula (NH4OOCCH2)2NCH2CH2N(CH2COOH)2
MDL Number MFCD00036454
UN Number NONH for all modes of transport
Beilstein 7785165
Synonym ethylenediaminetetraacetic acid diammonium salt hydrate, edetic acid diamine hydrate, diazanium,2-2-bis carboxymethyl amino ethyl-carboxylatome, ammonium 2,2-2-bis carboxymethyl amino ethyl azanediyl diacetate hydrate
InChI Key WRSUMHYBOYWMTD-UHFFFAOYSA-N
SMILES C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.N.N.O
IUPAC Name azane;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;hydrate
Molecular Weight (g/mol) 344.321
PubChem CID 71311375
Formula Weight 326.3g/mol
Packaging Poly Bottle
Chemical Name or Material Ethylenediaminetetraacetic Acid Diammonium Salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.