missing translation for 'onlineSavingsMsg'
Learn More

EDR1/PHC1 Antibody, Novus Biologicals™

Product Code. 18382098 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18382098 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18382098 Supplier Novus Biologicals Supplier No. H00001911B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

EDR1/PHC1 Polyclonal antibody specifically detects EDR1/PHC1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen EDR1/PHC1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. AAH73964
Gene Alias early development regulator 1 (homolog of polyhomeotic 1), Early development regulatory protein 1, EDR1, hPH1, PH1, polyhomeotic homolog 1 (Drosophila), polyhomeotic-like 1, polyhomeotic-like 1 (Drosophila), polyhomeotic-like protein 1, RAE28
Host Species Mouse
Immunogen PHC1 (AAH73964, 1 a.a. - 957 a.a.) full-length human protein. METESEQNSNSTNGSSSSGGSSRPQIAQMSLYERQAVQATIAASRQASSPNTSTTQQQTTTTQASINLATTSAAQLISRSQSVSSPSATTLTQSVLLGNTTSPPLNQSQAQMYLRVNRTLGRNVPLASQLILMPNGAVAAVQQEVPSAQSPGVHADADQVQNLAVRNQQASAQGPQMQGSTQKAIPPGASPVSSLSQASSQALAVAQASSGATNQSLNLSQAGGGSGNSIPGSMGPGGGGQAHGGLGQLPSSGMGGGSCPRKGTGVVQPLPAAQTVTVSQGSQTEAESAAAKKAEADGSGQQNVGMNLTRTATPAPSQTLISSATYTQIQPHSLIQQQQQIHLQQKQVVIQQQIAIHHQQQFQHRQSQLLHTATHLQLAQQQQQQQQQQQQQPQATTLTAPQPPQVPPTQQVPPSQSQQQAQTLVVQPMLQSSPLSLPPDAAPKPPIPIQSKPPVAPIKPPQLGAAKMSAAQQPPPHIPVQVVGTRQPGTAQAQALGLAQLAAAVPTSRGMPGTVQSGQAHLASSPPSSQAPGALQECPPTLAPGMTLAPVQGTAHVVKGGATTSSPVVAQVPAAFYMQSVHLPGKPQTLAVKRKADSEEERDDVSTLGSMLPAKASPVAESPKVMDEKSSLGEKAESVANVNANTPSSELVALTPAPSVPPPTLAMVSRQMGDSKPPQAIVKPQILTHIIEGFVIQEGAEPFPVGCSQLLKESEKPLQTGLPTGLTENQSGGSLGVDSPSAELDKKANLLKCEYCGKYAPAEQFRGSKRFCSMTCAKRYNVSCSHQFRLKRKKMKEFQEANYARVRRRGPRRSSSDIARAKIQGKCHRGQEDSSRGSDNSSYDEALSPTSPGPLSVRAGHGERDLGNPNTAPPTPELHGINPVFLSSNPSRWSVEEVYEFIASLQGCQEIAEEFRSQEIDGQALLLLKEEHLMSAMNIKLGPALKICAKINVLKET
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1911
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.