missing translation for 'onlineSavingsMsg'
Learn More

Edoxudine, MedChemExpress

Product Code. 16494517
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16494517

Brand: Medchem Express HYB1011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Edoxudine is an antiviral drug, is an analog of thymidine, shows effectiveness against herpes simplex virus.

Edoxudine is an antiviral drug, is an analog of thymidine, shows effectiveness against herpes simplex virus.

Specifications

Chemical Name or Material Edoxudine
Physical Form Solid
CAS 15176-29-1
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C11H16N2O5
For Use With (Application) COVID-19-anti-virus
Synonym EUDR
Solubility Information DMSO : ≥ 125 mg/mL (487.79 mM)
SMILES OC[C@@H]1[C@H](C[C@H](N2C(NC(C(CC)=C2)=O)=O)O1)O
Molecular Weight (g/mol) 256.26
Formula Weight 256.26
Percent Purity 98.56%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.