missing translation for 'onlineSavingsMsg'
Learn More

Edoxaban Tosylate Monohydrate, TRC

Product Code. 30006194
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006194 1 g 1g
30007688 100 mg 100mg
30008237 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006194 Supplier Toronto Research Chemicals Supplier No. TRCE5555251G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1229194-11-9
Molecular Formula C31H38ClN7O7S2•xH2O
Molecular Weight (g/mol) 720.26
Synonym LIXIANA, N-(5-Chloropyridin-2-yl)-N'-((1S,2R,4S)-4-[(dimethyl amino)carbonyl]-2-[[(5-methyl-4,5,6,7-tetrahydrothiazolo[5,4-c]pyridin-2-yl)carbonyl]amino]cyclohexyl)ethanediamide p-Toluenesulfonate hydrate N-(5-Chloropyridin-2-yl)-N'-[(1S,2R,4S)-4-[(dimethylamino)carbonyl]-2-[[(5-methyl-4,5,6,7-tetrahydrothiazolo[5,4-c]pyridin-2-yl)carbonyl]amino]cyclohexyl]ethanediamide mono(p-toluenesulfonate) hydrate
SMILES O=C(C1=NC(CCN(C)C2)=C2S1)N[C@H](C[C@@H](C(N(C)C)=O)CC3)[C@H]3NC(C(NC4=CC=C(Cl)C=N4)=O)=O.OS(C5=CC=C(C)C=C5)(=O)=O

Specifications

InChI Formula InChI=1S/C24H30ClN7O4S.C7H8O3S.H2O/c1-31(2)24(36)13-4-6-15(27-20(33)21(34)30-19-7-5-14(25)11-26-19)17(10-13)28-22(35)23-29-16-8-9-32(3)12-18(16)37-23;1-6-2-4-7(5-3-6)11(8,9)10;/h5,7,11,13,15,17H,4,6,8-10,12H2,1-3H3,(H,27,33)(H,28,35)(H,26,30,34);2-5H,1H3,(H,8,9,10);1H2/t13-,15-,17+;;/m0../s1
Quantity 1 g
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material Edoxaban Tosylate Hydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.