missing translation for 'onlineSavingsMsg'
Learn More

Edoxaban tosylate monohydrate, MedChemExpress

Product Code. 16476477
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476477 5 mg 5mg
16474816 100 mg 100mg
16450137 50 mg 50mg
16413758 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16476477 Supplier Medchem Express Supplier No. HY10264B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Edoxaban (DU-176b) monohydrate is an orally active, highly potent, selective, and direct Factor Xa (FXa) inhibitor with Kis of 0.561 and 2.98 nM for free human FXa and prothrombinase. Edoxaban monohydrate exhibits more than 10,000-fold selectivity over other coagulation proteases. Edoxaban monohydrate can be used for preventing thromboembolic disease research.

Edoxaban (DU-176b) monohydrate is an orally active, highly potent, selective, and direct Factor Xa (FXa) inhibitor with Kis of 0.561 and 2.98 nM for free human FXa and prothrombinase. Edoxaban monohydrate exhibits more than 10,000-fold selectivity over other coagulation proteases. Edoxaban monohydrate can be used for preventing thromboembolic disease research.

Specifications

Chemical Name or Material Edoxaban tosylate monohydrate
CAS 1229194-11-9
Color White
Purity Grade Notes Research
Molecular Formula C31H40ClN7O8S2
Quantity 5 mg
Synonym DU-176b monohydrate
Solubility Information DMSO : 50 mg/mL (67.73 mM; Need ultrasonic) ∣H2O : 1 mg/mL (1.35 mM; Need ultrasonic)
SMILES O=C(N(C)C)[C@@H](CC1)C[C@H]([C@H]1NC(C(NC(C=C2)=NC=C2Cl)=O)=O)NC(C3=NC(CC4)=C(CN4C)S3)=O.O=S(C5=CC=C(C)C=C5)(O)=O.O
Molecular Weight (g/mol) 738.27
Formula Weight 738.27
Percent Purity 98.3%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.