missing translation for 'onlineSavingsMsg'
Learn More

EDO-S101, MedChemExpress

Product Code. 16496287
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496287 5 mg 5mg
16469976 50 mg 50mg
16464676 100 mg 100mg
16462776 25 mg 25mg
16453568 10 mg 10mg
16400909 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16496287 Supplier Medchem Express Supplier No. HY101780
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress EDO-S101 (Tinostamustine) is a pan HDAC inhibitor; inhibits HDAC6, HDAC1, HDAC2 and HDAC3 with IC50 values of 6 nM, 9 nM, 9 nM and 25 nM, respectively.

EDO-S101 (Tinostamustine) is a pan HDAC inhibitor

Specifications

Chemical Name or Material EDO-S101
CAS 1236199-60-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H28Cl2N4O2
Quantity 5 mg
Synonym Tinostamustine
Solubility Information DMSO : 100 mg/mL (240.76 mM; Need ultrasonic)
SMILES O=C(NO)CCCCCCC1=NC2=CC(N(CCCl)CCCl)=CC=C2N1C
Molecular Weight (g/mol) 415.36
Formula Weight 415.36
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.