missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDIL3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12580-1-AP-150UL
This item is not returnable.
View return policy
Description
Del-1 is an integrin ligand that is expressed by endothelial cells during embryological vascular development. It promotes adhesion of endothelial cells through an interaction with the alpha-v/beta-3 integrin receptor. Del-1 plays a vital role in mediating angiogenesis.Specifications
| EDIL3 | |
| Polyclonal | |
| Unconjugated | |
| EDIL3 | |
| DEL1, EDIL3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10085, 13612, 688400 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O35474, O43854 | |
| Edil3 | |
| EDIL3 Fusion Protein Ag3274 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction