missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EDIL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522998
This item is not returnable.
View return policy
Description
Suggested positive control: normal human brain lysate, normal mouse brain lysate.
Del-1 is an integrin ligand that is expressed by endothelial cells during embryological vascular development. It promotes adhesion of endothelial cells through an interaction with the alpha-v/beta-3 integrin receptor. Del-1 plays a vital role in mediating angiogenesis.
Specifications
| EDIL3 | |
| Polyclonal | |
| Unconjugated | |
| EDIL3 | |
| Del1; Del-1; developmental endothelial locus-1; developmentally-regulated endothelial cell locus 1 protein; Edil3; EGF like repeats and discoidin domains 3; EGF-like repeat and discoidin I-like domain-containing protein 3; EGF-like repeats and discoidin I-like domains 3; integrin-binding protein DEL1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10085, 13612 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1.62 mg/mL | |
| PBS with 30% glycerol and 0.1% sodium azide | |
| O35474, O43854 | |
| EDIL3 | |
| Synthetic peptide made to an internal portion of human Del-1 (within residues 200-350). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction