missing translation for 'onlineSavingsMsg'
Learn More

EDIL3/DEL1 Antibody, Novus Biologicals™

Product Code. 18328069 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18328069 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18328069 Supplier Novus Biologicals Supplier No. H00010085B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

EDIL3/DEL1 Polyclonal antibody specifically detects EDIL3/DEL1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen EDIL3/DEL1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunocytochemistry/ Immunofluorescence
Formulation PBS (pH 7.4)
Gene Accession No. NP_005702.3
Gene Alias DEL1developmental endothelial locus-1, Developmentally-regulated endothelial cell locus 1 protein, EGF-like repeat and discoidin I-like domain-containing protein 3, EGF-like repeats and discoidin I-like domains 3, Integrin-binding protein DEL1, MGC26287
Host Species Mouse
Immunogen EDIL3 (NP_005702.3, 1 a.a. - 480 a.a.) full-length human protein. MKRSVAVWLLVGLSLGVPQFGKGDICDPNPCENGGICLPGLADGSFSCECPDGFTDPNCSSVVEVASDEEEPTSAGPCTPNPCHNGGTCEISEAYRGDTFIGYVCKCPRGFNGIHCQHNINECEVEPCKNGGICTDLVANYSCECPGEFMGRNCQYKCSGPLGIEGGIISNQQITASSTHRALFGLQKWYPYYARLNKKGLINAWTAAENDRWPWIQINLQRKMRVTGVITQGAKRIGSPEYIKSYKIAYSNDGKTWAMYKVKGTNEDMVFRGNIDNNTPYANSFTPPIKAQYVRLYPQVCRRHCTLRMELLGCELSGCSEPLGMKSGHIQDYQITASSIFRTLNMDMFTWEPRKARLDKQGKVNAWTSGHNDQSQWLQVDLLVPTKVTGIITQGAKDFGHVQFVGSYKLAYSNDGEHWTVYQDEKQRKDKVFQGNFDNDTHRKNVIDPPIYARHIRILPWSWYGRITLRSELLGCTEEE
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Angiogenesis, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10085
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.