missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
EDIL3/DEL1 Polyclonal antibody specifically detects EDIL3/DEL1 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | EDIL3/DEL1 |
| Applications | Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | DyLight 680 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DEL1developmental endothelial locus-1, Developmentally-regulated endothelial cell locus 1 protein, EGF-like repeat and discoidin I-like domain-containing protein 3, EGF-like repeats and discoidin I-like domains 3, Integrin-binding protein DEL1, MGC26287 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide made to an internal portion of human Del-1 (within residues 200-350). [Swiss-Prot# O43854] |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?