missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDIL3/DEL1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£170.00 - £405.00
Specifications
| Antigen | EDIL3/DEL1 |
|---|---|
| Concentration | 1.62 mg/mL |
| Dilution | Western Blot 2 ug/ml, Simple Western 1:20, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18016374
|
Novus Biologicals
NBP1-28632SS |
0.025 mL |
£170.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18794963
|
Novus Biologicals
NBP1-28632 |
0.1 mL |
£405.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EDIL3/DEL1 Polyclonal specifically detects EDIL3/DEL1 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence.Specifications
| EDIL3/DEL1 | |
| Western Blot 2 ug/ml, Simple Western 1:20, Immunocytochemistry/Immunofluorescence 1:100 | |
| Polyclonal | |
| Rabbit | |
| Angiogenesis, Cytoskeleton Markers, Endothelial Cell Markers, Extracellular Matrix, Signal Transduction | |
| PBS and 30% Glycerol with 0.1% Sodium Azide | |
| DEL1developmental endothelial locus-1, Developmentally-regulated endothelial cell locus 1 protein, EGF-like repeat and discoidin I-like domain-containing protein 3, EGF-like repeats and discoidin I-like domains 3, Integrin-binding protein DEL1, MGC26287 | |
| EDIL3 | |
| IgG | |
| Affinity Purified | |
| 53 kDa |
| 1.62 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| O43854 | |
| 10085 | |
| Synthetic peptide made to an internal portion of human Del-1 (within residues 200-350). [Swiss-Prot# O43854] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title