missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDIL3/DEL1 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-28632AF350
This item is not returnable.
View return policy
Description
EDIL3/DEL1 Polyclonal specifically detects EDIL3/DEL1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| EDIL3/DEL1 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| DEL1developmental endothelial locus-1, Developmentally-regulated endothelial cell locus 1 protein, EGF-like repeat and discoidin I-like domain-containing protein 3, EGF-like repeats and discoidin I-like domains 3, Integrin-binding protein DEL1, MGC26287 | |
| Rabbit | |
| Affinity Purified | |
| Angiogenesis, Cytoskeleton Markers, Endothelial Cell Markers, Extracellular Matrix, Signal Transduction | |
| 10085.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| EDIL3 | |
| Synthetic peptide made to an internal portion of human Del-1 (within residues 200-350). [Swiss-ProtNo. O43854] | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction