missing translation for 'onlineSavingsMsg'
Learn More

Edicotinib, MedChemExpress

Product Code. 16493016
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493016 25 mg 25mg
16436997 5 mg 5mg
16430607 50 mg 50mg
16425206 100 mg 100mg
16414278 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16493016 Supplier Medchem Express Supplier No. HY109086

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Edicotinib (JNJ-40346527) is a potent, selective, brain penetrant and orally active colony-stimulating factor-1 receptor (CSF-1R) inhibitor with an IC50 of 3.2 nM.

Edicotinib exhibits less inhibitory effects on KIT and FLT3 with IC50 values of 20 nM and 190 nM, respectively. Edicotinib limits microglial expansion and attenuates microglial proliferation and neurodegeneration in mice. Edicotinib has the potential for Alzheimer’s disease and rheumatoid arthritis research.

Specifications

Chemical Name or Material Edicotinib
CAS 1142363-52-7
Color White
Purity Grade Notes Research
Molecular Formula C27H35N5O2
Quantity 25 mg
Synonym JNJ-40346527 JNJ-527
Solubility Information DMSO : 16.67 mg/mL (36.11 mM; Need ultrasonic)
SMILES O=C(C1=NC=C(C#N)N1)NC2=C(C3=CCC(C)(C)CC3)N=C(C(CC(C)(C)O4)CC4(C)C)C=C2
Molecular Weight (g/mol) 461.6
Formula Weight 461.6
Percent Purity 98.14%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.