Learn More
Epredia™ Edge-Rite™ Disposable Microtome Blades
Product Code. 12729346 Shop All Epredia Products
Click to view available options
Profile:
High
Low
Unit Size:
Pack of 50
Product Code. 12729346
Brand: Epredia™ 4280L
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Experience consistent quality from blade to blade and sample to sample with Epredia™ Edge-Rite™ disposable microtome blades.
  • Surgical-grade steel
  • High- and low-profile blades are specially coated to increase longevity and optimize section quality
Low profile blades:
  • Blades are 20% thicker than traditional blades to increase knife edge stability, which allows for excellent ribboning and facilitates usage in cryostats
  • Blades are 5% longer, allowing for more sections per blade
  • Blades fit any low-profile blade holder
TRUSTED_SUSTAINABILITY
Specifications
Product Type Disposable Microtome Blade
Profile Low
Material Surgical-grade Steel
Disposable Single-use