missing translation for 'onlineSavingsMsg'
Learn More

Ansell EDGE™ 48-919 Double Layer Nitrile Coated Oil Repellent Gloves

Product Code. 17687455 Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
11
12
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 17687455

Brand: Ansell 48919060

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Double layer nitrile coating keeps hands dry and comfortable when handling oily parts, while an elastic knit wrist ensures a snug fit.

  • Seamless polyester liner is comfortable and flexible
  • Double nitrile coating provides protection against industrial fluid exposure
  • Rough texture on the palm ensures a secure grip in oily environments
  • Ideal for workers who handle small and medium-sized components coated with oil or lubricant
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Coating Material Nitrile
Color Red, Black
Exterior Finish Fully Textured
Hand Specific
Latex Free Yes
Material Polyester
Product Type Dip-coated Gloves
Size 6
For Use With (Application) Automotive, Construction, Machinery/Equipment, and Metal Fabrication
Glove Cuff Color Gray
Glove Cuff Style Knit
Series 48-919
Yarn Gauge 13
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.