Learn More
Ansell EDGE™ 48-701 Protective Gloves
Product Code. 18896956 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
11
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size
18896956 8
18886956 7
18806966 9
18816966 10
18826966 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 18896956 Supplier Ansell Supplier No. 48701080
  • ONLINE EXCLUSIVE PRICE
    £72.60
    £63.73 / 12 pairs
    Save £8.87
    12% Off
Estimated Shipment: 15-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Cut, oil and abrasion-resistant gloves, granting good grip in dry conditions.
  • Optimized durability: EDGE™ 48-701 industrial gloves' palm PU coating provides a high level of abrasion protection (ANSI 6), moderate cut resistance (ANSI A2), a good grip in dry environments, and better protection against oil than knitted or cut and sewn safety work gloves
  • Increased protection: EDGE 48-701 protective gloves provide two levels of cut performance, affording robust, all-round hand protection in mediumweight applications
  • Enhanced features: A more affordable alternative to medium-duty leather, thick cotton or aramid gloves, EDGE™ 48-701 high-quality work gloves offer a seamless liner and knitted wrist, which deliver form-fitting comfort, good dexterity and added wrist protection
TRUSTED_SUSTAINABILITY
Specifications
Coating Material Polyurethane
Color Gray
Cut Resistance Rating Level B
Hand Specific
Latex Free Yes
Material Spandex, Polyester, Polyethylene, Polyurethane
Product Type Industrial Gloves
Size 8