missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EDG6 Monoclonal Antibody (1), FITC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA112718
This item is not returnable.
View return policy
Description
EDG6 Monoclonal Antibody for Western Blot, Flow
EDG6 (S1P4) belongs to a family of G-protein coupled receptors whose ligands are lysophospholipids. The ligand for EDG6 (S1P4) is sphingosine-1-phosphate. There are 8 known members of the EDG receptor family and they are implicated in mediating growth related effects such as induction of cellular proliferation, alterations in differentiation and survival and suppression of apoptosis. They also evoke cellular effector functions that are dependent on cytoskeletal responses such as contraction, secretion, adhesion and chemotaxis. EDG receptors are developmentally regulated and differ in tissue distribution. They couple to multiple types of G proteins to signal through ras and MAP kinase, rho, phospholipase C and several protein tyrosine kinases. EDG6 (S1P4) is expressed in lymphoid and hematopoietic tissue and in the lungs. Recently, the designation of the EDG receptors has been modified to include the ligand for the receptor. Thus, the new designation for the EDG6 (S1P4) receptor is the sphingosine 1-phosphate receptor-4 (S1P4).
Specifications
| EDG6 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.08% sodium azide | |
| O95977 | |
| S1PR4 | |
| Synthetic peptide corresponding to the C-terminal of EDG6. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| 1 | |
| FITC | |
| S1PR4 | |
| Edg6; EDG-6; Endothelial differentiation G-protein coupled receptor 6; endothelial differentiation G-protein-coupled receptor 6; endothelial differentiation, G protein coupled receptor 6; endothelial differentiation, G-protein-coupled receptor 6; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor 6; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor, 6; LPC1; Lysophospholipid receptor C1; S1P receptor 4; S1P receptor Edg-6; S1P(4) receptor; S1p4; S1PR4; SLP4; sphingosine 1-phosphate receptor 4; Sphingosine 1-phosphate receptor Edg-6; sphingosine-1-phosphate receptor 4 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 8698 | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction