missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDG4 Polyclonal Antibody

EDG4 Polyclonal Antibody
Brand: Invitrogen PA550586
This item is not returnable.
View return policy
Description
EDG4 Polyclonal Antibody for IHC (P)
Specifications
| EDG4 | |
| Polyclonal | |
| Unconjugated | |
| LPAR2 | |
| Edg4; EDG-4; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor 4; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor, 4; G protein-coupled receptor; IPA2; LPA receptor 2; LPA receptor EDG4; LPA2; LPA-2; LPA2 Receptor; LPAR2; Lysophosphatidic acid receptor 2; lysophosphatidic acid receptor EDG4; lysophosphatidic acid receptor Edg-4; Pbx4; pre-B-cell leukemia transcription factor 4; RGD1561336 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9170 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.6 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9HBW0 | |
| LPAR2 | |
| Synthetic peptide corresponding to residues near the C terminal of human lysophosphatidic acid receptor 2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction