missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDF1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12419-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene is known to suppress G-protein and mitogen-activated signal transduction in mammalian cells. The encoded protein shares significant similarity with Arabidopsis FUS6, which is a regulator of light-mediated signal transduction in plant cells. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| EDF1 | |
| Polyclonal | |
| Unconjugated | |
| EDF1 | |
| EDF 1, EDF1, MBF1, Multiprotein bridging factor 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 59022, 8721 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O60869, Q9JMG1 | |
| Edf1 | |
| EDF1 Fusion Protein Ag3063 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction