missing translation for 'onlineSavingsMsg'
Learn More

EDF1 Antibody, DyLight 405, Novus Biologicals™

Product Code. 18425237 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425237 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425237 Supplier Novus Biologicals Supplier No. NBP222327V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EDF1 Polyclonal antibody specifically detects EDF1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen EDF1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EDF-1Multiprotein-bridging factor 1, endothelial differentiation-related factor 1, MBF1, MGC9058, multiprotein bridging factor 1, multiprotein bridging factor-1
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 98 to 148 of human Endothelial Differentiation-Related Factor 1 using the numbering given in entry NP_003783.1 (GeneID 8721).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8721
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.