missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDEM1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA556191
This item is not returnable.
View return policy
Description
Immunogen sequence: SLEKYTKVKC GYATLHHVID KSTEDRMESF FLSETCKYLY LLFDEDNPVH KSGTRYMFTT EGHIVSVDEH LRELPWKEFF SEEGGQDQGG KSVHRPKPHE LKVINSSSNC NRVPDE Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.
EDEM1 belongs to the glycosyl hydrolase 47 family. It extracts misfolded glycoproteins, but not glycoproteins undergoing productive folding, from the calnexin cycle. It is directly involved in endoplasmic reticulum-associated degradation (ERAD) and targets misfolded glycoproteins for degradation in an N-glycan-dependent manner.
Specifications
| EDEM1 | |
| Polyclonal | |
| Unconjugated | |
| EDEM1 | |
| A130059K23Rik; EDEM; EDEM1; ER degradation enhancer, mannosidase alpha-like 1; ER degradation enhancing alpha-mannosidase like protein 1; ER degradation ER degradation enhancer, mannosidase alpha-like 1; ER degradation-enhancing alpha-mannosidase-like 1; ER degradation-enhancing alpha-mannosidase-like protein 1; KIAA0212; mKIAA0212; RGD1563633 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9695 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q92611 | |
| EDEM1 | |
| Recombinant protein corresponding to Human EDEM1 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur