missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EDC4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530485
This item is not returnable.
View return policy
Description
Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7, mouse brain. Predicted reactivity: Mouse (96%), Rat (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
In the process of mRNA degradation, seems to play a role in mRNA decapping. Component of a complex containing DCP2 and DCP1A which functions in decapping of ARE-containing mRNAs. Promotes complex formation between DCP1A and DCP2. Enhances the catalytic activity of DCP2 (in vitro).
Specifications
| EDC4 | |
| Polyclonal | |
| Unconjugated | |
| EDC4 | |
| Autoantigen Ge-1; autoantigen RCD8; autoantigen RCD-8; BC022641; Edc4; enhancer of mRNA decapping 4; enhancer of mRNA-decapping protein 4; fe26b05; GE1; Ge-1; HEDL5; HEDLS; Human enhancer of decapping large subunit; LRRG00115; Nrn1l; RCD8; RCD-8; RGD1309136; si:ch211-237c7.1; si:dkey-16n13.6; wu:fe26b05 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 234699, 23644 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q3UJB9, Q6P2E9 | |
| EDC4 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human EDC4. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction