missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDC4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-2273
This item is not returnable.
View return policy
Description
EDC4 Polyclonal specifically detects EDC4 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| EDC4 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| Autoantigen Ge-1, Autoantigen RCD-8, enhancer of mRNA decapping 4, enhancer of mRNA-decapping protein 4, Ge-1, Hedls, HEDLSHuman enhancer of decapping large subunit, RCD-8 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 23644 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot, Immunoprecipitation 2-5 ug/mg lysate | |
| Q6P2E9 | |
| EDC4 | |
| The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human RCD8 Autoantigen using the numbering given in entry AAH43616 (GeneID 23644). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction