missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EDC3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522395
This item is not returnable.
View return policy
Description
Recommended positive controls: A431, NIH-3T3, C2C12. Predicted reactivity: Mouse (95%), Xenopus laevis (82%), Chicken (92%), Rhesus Monkey (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein that is important in mRNA degradation. The encoded protein is a component of a decapping complex that promotes efficient removal of the monomethylguanosine (m7G) cap from mRNAs, as part of the 5' to 3' mRNA decay pathway. Mutations in this gene have been identified in human patients with an autosomal recessive form of intellectual disability.
Specifications
| EDC3 | |
| Polyclonal | |
| Unconjugated | |
| Edc3 | |
| AA517853; EDC3; enhancer of mRNA decapping 3; enhancer of mRNA decapping 3 homolog; enhancer of mRNA decapping 3 homolog (S. cerevisiae); enhancer of mRNA-decapping protein 3; hYjeF_N2; hYjeF_N2-15q23; hypothetical protein FLJ21128; hypothetical protein LOC504054; im:7151288; Lsm16; LSM16 homolog; LSM16 homolog (EDC3, S. cerevisiae); LSM16 protein homolog; MRT50; PP844; RGD1306898; wu:fb82d07; Yjdc; yjeF domain containing; yjeF domain-containing protein 1; YjeF N-terminal domain-containing protein 2; yjeF_N2; YJEFN2; zgc:112006 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 353190, 80153 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8K2D3, Q96F86 | |
| Edc3 | |
| Recombinant protein encompassing a sequence within the center region of human Edc3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction