missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EDC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116422
This item is not returnable.
View return policy
Description
EDC3 Polyclonal Antibody for Western Blot
This gene encodes a protein that is important in mRNA degradation. The encoded protein is a component of a decapping complex that promotes efficient removal of the monomethylguanosine (m7G) cap from mRNAs, as part of the 5' to 3' mRNA decay pathway. Mutations in this gene have been identified in human patients with an autosomal recessive form of intellectual disability.
Specifications
| EDC3 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q502M5 | |
| Edc3 | |
| KLH conjugated synthetic peptide between 162-197 amino acids from the Central region of (DANRE) edc3. | |
| 400 μL | |
| Primary | |
| Zebrafish | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Edc3 | |
| AA517853; EDC3; enhancer of mRNA decapping 3; enhancer of mRNA decapping 3 homolog; enhancer of mRNA decapping 3 homolog (S. cerevisiae); enhancer of mRNA-decapping protein 3; hYjeF_N2; hYjeF_N2-15q23; hypothetical protein FLJ21128; hypothetical protein LOC504054; im:7151288; Lsm16; LSM16 homolog; LSM16 homolog (EDC3, S. cerevisiae); LSM16 protein homolog; MRT50; PP844; RGD1306898; wu:fb82d07; Yjdc; yjeF domain containing; yjeF domain-containing protein 1; YjeF N-terminal domain-containing protein 2; yjeF_N2; YJEFN2; zgc:112006 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 504054 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction