missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EDAR Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA596290

Product Code. 16539131

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: EYSNCGENEY YNQTTGLCQE CPPCGPGEEP YLSCGYGTKD EDYGCVPCPA EKFSKGGYQI CRRHKDCEGF FRATVLTPGD MENDAECGPC LPGYYMLENR PRNIYGMVCY SCLLAPPNTK ECVGATSGAS ANFPGTSGSS TLSPFQHAHK ELSGQGHLAT A; Positive Samples: 293T, LO2, SW480; Cellular Location: Membrane, Single-pass type I membrane protein.

This gene encodes a member of the tumor necrosis factor receptor family. The encoded transmembrane protein is a receptor for the soluble ligand ectodysplasin A, and can activate the nuclear factor-kappaB, JNK, and caspase-independent cell death pathways. It is required for the development of hair, teeth, and other ectodermal derivatives. Mutations in this gene result in autosomal dominant and recessive forms of hypohidrotic ectodermal dysplasia.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EDAR
Polyclonal
Unconjugated
Edar
Anhidrotic ectodysplasin receptor 1; DL; Downless; downless homolog; downless, mouse, homolog of; ECTD10A; ECTD10B; ectodermal dysplasia receptor; ectodysplasin 1, anhidrotic receptor; ectodysplasin A receptor; ectodysplasin-A receptor; ED1R; ED3; ED5; EDA1R; EDA3; EDA-A1 receptor; EDA-A1R; Edar; HRM1; RGD1561714; Tumor necrosis factor receptor superfamily member EDAR
Rabbit
Affinity Chromatography
RUO
10913
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.88 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UNE0
Edar
Recombinant fusion protein containing a sequence corresponding to amino acids 27-187 of human EDAR (NP_0717311).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.