missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EDAR Polyclonal Antibody
GREENER_CHOICE

Product Code. 15787781 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15787781 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15787781 Supplier Invitrogen™ Supplier No. PA559938

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: TKDEDYGCVP CPAEKFSKGG YQICRRHKDC EGFFRATVLT PGDMENDAEC GPCLPGYYML ENRPRNIYGM VCYSCLLAPP NTKECVGATS GASANFPGTS Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 95%.

This gene encodes a member of the tumor necrosis factor receptor family. The encoded transmembrane protein is a receptor for the soluble ligand ectodysplasin A, and can activate the nuclear factor-kappaB, JNK, and caspase-independent cell death pathways. It is required for the development of hair, teeth, and other ectodermal derivatives. Mutations in this gene result in autosomal dominant and recessive forms of hypohidrotic ectodermal dysplasia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EDAR
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Edar
Gene Accession No. Q9UNE0
Gene Alias Anhidrotic ectodysplasin receptor 1; DL; Downless; downless homolog; downless, mouse, homolog of; ECTD10A; ECTD10B; ectodermal dysplasia receptor; ectodysplasin 1, anhidrotic receptor; ectodysplasin A receptor; ectodysplasin-A receptor; ED1R; ED3; ED5; EDA1R; EDA3; EDA-A1 receptor; EDA-A1R; Edar; HRM1; RGD1561714; Tumor necrosis factor receptor superfamily member EDAR
Gene Symbols Edar
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EDAR. Recombinant protein control fragment (Product #RP-90259).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10913
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.