missing translation for 'onlineSavingsMsg'
Learn More

EDAR Antibody (EDAR12), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18427746 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427746 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427746 Supplier Novus Biologicals Supplier No. NBP281103JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

EDAR Monoclonal antibody specifically detects EDAR in Human, Mouse, Rat, Canine, Chicken samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen EDAR
Applications Western Blot, ELISA, Immunocytochemistry, Neutralization
Classification Monoclonal
Clone EDAR12
Conjugate Janelia Fluor 549
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Surface Plasmon Resonance, Block/Neutralize
Formulation 50mM Sodium Borate
Gene Alias Anhidrotic ectodysplasin receptor 1, DLED3, Ectodermal dysplasia receptor, ectodysplasin 1, anhidrotic receptor, ectodysplasin A receptor, Ectodysplasin-A receptor, ED1R, ED5, EDA1R, EDA3EDA-A1 receptor, EDA-A1R, Edar, FLJ94390, HRM1, mouse, homolog of, tumor necrosis factor receptor superfamily member EDAR
Host Species Rabbit
Immunogen EDAR12 was prepared by immunixzing female OVE1B mice (with the Edar gene deleted) subcutaneously with mouse EDAR-Fc and positive hybridoma clones were screened for binding mEDAR by ELISA.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 10913
Target Species Human, Mouse, Rat, Canine, Chicken
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.