missing translation for 'onlineSavingsMsg'
Learn More

EDAR Antibody (EDAR12), DyLight 405, Novus Biologicals™

Product Code. 18397820 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18397820 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18397820 Supplier Novus Biologicals Supplier No. NBP281103V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

EDAR Monoclonal specifically detects EDAR in Human, Mouse, Rat, Canine, Chicken samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EDAR
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone EDAR12
Conjugate DyLight 405
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Anhidrotic ectodysplasin receptor 1, DLED3, Ectodermal dysplasia receptor, ectodysplasin 1, anhidrotic receptor, ectodysplasin A receptor, Ectodysplasin-A receptor, ED1R, ED5, EDA1R, EDA3EDA-A1 receptor, EDA-A1R, Edar, FLJ94390, HRM1, mouse, homolog of, tumor necrosis factor receptor superfamily member EDAR
Gene Symbols EDAR
Host Species Rabbit
Immunogen EDAR12 was prepared by immunixzing female OVE1B mice (with the Edar gene deleted) subcutaneously with mouse EDAR-Fc and positive hybridoma clones were screened for binding mEDAR by ELISA.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 10913
Test Specificity EDAR12 recognises and binds to CRD1 of the extracellular domain of mouse EDAR. EDAR3 cross-reacts with EDAR derived from human, dog, rat and chicken when EDAR is fused to the glycosylphosphatidylinositol anchor of TRAILR3. EDAR is the receptor for the TNF family ligand EDA1, which is a type II transmembrane protein possessing a collagen-like domain and a C-terminal TNF-homology domain. EDAR is important for the correct development of skin appendages including hair, teeth and eccrine sweat glands. LoF mutations in the Eda gene is known to cause XLHED (X-linked hypohidrotic ectodermal dysplasia), and results in abnormal development.
Target Species Human, Mouse, Rat, Canine, Chicken
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.