missing translation for 'onlineSavingsMsg'
Learn More

EDA2R/TNFRSF27/XEDAR Antibody, DyLight 488, Novus Biologicals™

Product Code. 18402768 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402768 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18402768 Supplier Novus Biologicals Supplier No. NBP176710G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EDA2R/TNFRSF27/XEDAR Polyclonal antibody specifically detects EDA2R/TNFRSF27/XEDAR in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen EDA2R/TNFRSF27/XEDAR
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias ectodysplasin A2 receptor, EDA-A2 receptor, EDA-A2R, EDAA2Rtumor necrosis factor receptor superfamily member XEDAR, EDAR2, XEDARTNFRSF27tumor necrosis factor receptor superfamily member 27, X-linked ectodysplasin-A2 receptor
Host Species Rabbit
Immunogen Antibody was raised against recombinant human XEDAR.
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 60401
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.