missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDA2R/TNFRSF27/XEDAR Antibody (072), Janelia Fluor™ 549, Novus Biologicals™
Shop All Bio Techne ProductsDescription
EDA2R/TNFRSF27/XEDAR Monoclonal antibody specifically detects EDA2R/TNFRSF27/XEDAR in Human samples. It is validated for ELISA
Specifications
Specifications
| Classification | Monoclonal |
| Clone | 072 |
| Gene Alias | ectodysplasin A2 receptor, EDA-A2 receptor, EDA-A2R, EDAA2Rtumor necrosis factor receptor superfamily member XEDAR, EDAR2, XEDARTNFRSF27tumor necrosis factor receptor superfamily member 27, X-linked ectodysplasin-A2 receptor |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 60401 |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?