missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EDA2R Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565878
This item is not returnable.
View return policy
Description
Immunogen sequence: CQENEYWDQWG RCVTCQRCGP GQELSKDCGY GEGGDAYCTA CPPRRYKSSW GHHKCQSCIT CAVINRVQKV NCTATS Highest antigen sequence identity to the following orthologs - mouse 86%, rat 88%.
X-linked ectodysplasin-A2 receptor (XEDAR) is a recently isolated member of the tumor necrosis factor receptor family that is highly expressed during embryonic development and binds to ectodysplatin-A2 (EDA-A2). Two predominantly expressed isoforms, XEDAR-s and XEDAR-L, differ by only a 21-amino region at the juxtamembrane region of the cytoplasmic domain. Neither isoform possesses a death domain and both have been shown to act mainly through TRAF3 and TRAF6 to activate the NF-kappa-B and JNK pathways. Cells transfected with XEDAR and treated with EDA-A2 cause the assembly of a secondary complex containing FADD, caspase-8 and caspase-10, leading to the activation caspase-8 and caspase-3, and finally apoptosis. The EDA-A2-induced apoptosis is dependent on caspase-9 activation, as various pharmacological and genetic inhibitors of caspase-8 blocked apoptosis following EDA-A2 treatment.
Specifications
| EDA2R | |
| Polyclonal | |
| Unconjugated | |
| EDA2R | |
| 9430060M22Rik; ectodysplasin A2 isoform receptor; ectodysplasin A2 receptor; EDA2R; EDA-A2 receptor; EDAA2R; EDA-A2R; RGD1564025; Tnfrsf27; Tumor necrosis factor receptor superfamily member 27; tumor necrosis factor receptor superfamily member XEDAR; UNQ2448/PRO5727/PRO34080; X linked ectodysplasin receptor; Xedar; X-linked ectodysplasin-A2 receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 60401 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HAV5 | |
| EDA2R | |
| Recombinant Human EDA2R. Recombinant protein control fragment (Product #RP-106549). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction