missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EDA2R Polyclonal Antibody
GREENER_CHOICE

Goat Polyclonal Antibody

Brand:  Invitrogen™ PA5148473

Product Code. 30535665

  • £415.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.2 mg/mL in sterile PBS.

X-linked ectodysplasin-A2 receptor (XEDAR) is a recently isolated member of the tumor necrosis factor receptor family that is highly expressed during embryonic development and binds to ectodysplatin-A2 (EDA-A2). Two predominantly expressed isoforms, XEDAR-s and XEDAR-L, differ by only a 21-amino region at the juxtamembrane region of the cytoplasmic domain. Neither isoform possesses a death domain and both have been shown to act mainly through TRAF3 and TRAF6 to activate the NF-kappa-B and JNK pathways. Cells transfected with XEDAR and treated with EDA-A2 cause the assembly of a secondary complex containing FADD, caspase-8 and caspase-10, leading to the activation caspase-8 and caspase-3, and finally apoptosis. The EDA-A2-induced apoptosis is dependent on caspase-9 activation, as various pharmacological and genetic inhibitors of caspase-8 blocked apoptosis following EDA-A2 treatment.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EDA2R
Polyclonal
Unconjugated
EDA2R
9430060M22Rik; ectodysplasin A2 isoform receptor; ectodysplasin A2 receptor; EDA2R; EDA-A2 receptor; EDAA2R; EDA-A2R; RGD1564025; Tnfrsf27; Tumor necrosis factor receptor superfamily member 27; tumor necrosis factor receptor superfamily member XEDAR; UNQ2448/PRO5727/PRO34080; X linked ectodysplasin receptor; Xedar; X-linked ectodysplasin-A2 receptor
Goat
Antigen affinity chromatography
RUO
60401
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Inhibition Assays, Western Blot
0.2 mg/mL
PBS with trehalose and no preservative
Q9HAV5
EDA2R
Mouse myeloma cell line NS0-derived recombinant human EDA2R/TNFRSF27/XEDAR, Met1-Glu136.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.