missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDA/Ectodysplasin Antibody (Renzo-2), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59772PE-0.1ML
This item is not returnable.
View return policy
Description
EDA/Ectodysplasin Monoclonal specifically detects EDA/Ectodysplasin in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| EDA/Ectodysplasin | |
| Monoclonal | |
| PE | |
| PBS | |
| EDA | |
| Recombinant human EDA-A1 (ectodysplasin-A1) extracellular domain (aa 245-391). | |
| 1896 | |
| Human, Mouse | |
| Purified |
| Flow Cytometry | |
| Renzo-2 | |
| Flow Cytometry | |
| ectodysplasin A, ED1 | |
| Mouse | |
| 0.1 mL | |
| Recognizes human and mouse EDA-A1 and EDA-A2. | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction