missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ectylurea, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Ectylurea
Specifications
Specifications
| Chemical Name or Material | Ectylurea |
| CAS | 95-04-5 |
| Analyte or Component Names | Ectylurea |
| Shipping Condition | Room Temperature |
| Molecular Formula | C7 H12 N2 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ectylurea, 2-Butenamide, N-(aminocarbonyl)-2-ethyl-, (2 Z)-, 2-Butenamide, N-(aminocarbonyl)-2-ethyl-, (Z)-, Urea, (2-ethylcrotonoyl)-, (Z)- (8 CI), (2-Ethyl-cis-crotonyl)urea, (alpha-Ethyl-cis-crotonyl)carbamide, Astyn, Cronil, Ectylcarbamide, Ektyl, Levanil, Nastyn, Neuroprocin, Nostal, Nostyn, Pacetyn |
| InChI Formula | InChI=1 S/C7H12N2O2/c1-3-5(4-2)6(10)9-7(8)11/h3H,4H2,1-2H3,(H3,8,9,10,11)/b5-3- |
| SMILES | CC\C(=C\C)\C(=O)NC(=O)N |
| IUPAC Name | (Z)-N-carbamoyl-2-ethylbut-2-enamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction