missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ecteinascidin 770, MedChemExpress
Description
Ecteinascidin 770 (ET-770) is a 1,2,3,4-tetrahydroisoquinoline alkaloid with potent anti-cancer activities
Specifications
Specifications
| Chemical Name or Material | Ecteinascidin 770 |
| CAS | 114899-80-8 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C40H42N4O10S |
| Quantity | 1 mg |
| Synonym | Ecteinascidine 770 Et-770 |
| Solubility Information | DMSO : 50 mg/mL (64.86 mM; Need ultrasonic) |
| SMILES | N#C[C@@H]([C@@]1([H])N(C)[C@]2([H])C3=C(C=C(C)C(OC)=C3O)C1)N4[C@@]2([H])[C@@](SC[C@@]5(NCC6)C7=C6C=C(O)C(OC)=C7)([H])C8=C(C(OCO9)=C9C(C)=C8OC(C)=O)[C@]4([H])COC5=O |
| Molecular Weight (g/mol) | 770.85 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?