missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ECT2L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548246
This item is not returnable.
View return policy
Description
ECT2L Polyclonal Antibody for Western Blot, IHC (P)
May act as a guanine nucleotide exchange factor (GEF).
Specifications
| ECT2L | |
| Polyclonal | |
| Unconjugated | |
| ECT2L | |
| ARHGEF32; C6orf91; dJ509I19.2; dJ509I19.3; dJ509I19.5; ECT2L; ECT2-like; epithelial cell transforming 2 like; epithelial cell transforming sequence 2 oncogene-like; epithelial cell-transforming sequence 2 oncogene-like; F-box protein 49; FBXO49; LFDH; lung specific F-box and DH domain containing protein; lung-specific F-box and DH domain-containing protein; Putative guanine nucleotide exchange factor LFDH | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 345930 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q008S8 | |
| ECT2L | |
| KLH conjugated synthetic peptide between 725-753 amino acids from the C-terminal region of human ECT2L. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction