missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ECT2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518166
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
The protein encoded by this gene is a transforming protein that is related to Rho-specific exchange factors and yeast cell cycle regulators. The expression of this gene is elevated with the onset of DNA synthesis and remains elevated during G2 and M phases. In situ hybridization analysis showed that expression is at a high level in cells undergoing mitosis in regenerating liver. Thus, this protein is expressed in a cell cycle-dependent manner during liver regeneration, and is thought to have an important role in the regulation of cytokinesis.
Specifications
| ECT2 | |
| Polyclonal | |
| Unconjugated | |
| ECT2 | |
| AI528536; ARHGEF31; Ect2; ect2 oncogene; ect2 protein; epithelial cell transforming 2; epithelial cell transforming sequence 2 oncogene; epithelial cell transforming sequence 2 oncogene protein; epithelial cell-transforming sequence 2 oncogene; FLJ10461; LOW QUALITY PROTEIN: protein ECT2; MGC138291; mKIAA4037; Protein ECT2; Protein ECT2-like protein | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 1894 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9H8V3 | |
| ECT2 | |
| Synthetic peptide sequence (AENSVLTSTTGRT) corresponding to the N-terminus amino acids of ECT2 (aa 2-15). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction