missing translation for 'onlineSavingsMsg'
Learn More

ECT2 Antibody, DyLight 755, Novus Biologicals™

Product Code. 18431707 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431707 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431707 Supplier Novus Biologicals Supplier No. NBP128729IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ECT2 Polyclonal antibody specifically detects ECT2 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ECT2
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias ARHGEF31, epithelial cell transforming sequence 2 oncogene, Epithelial cell-transforming sequence 2 oncogene, FLJ10461, MGC138291, protein ECT2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 833 and 883 of human epithelial cell transforming sequence 2 using the numbering given in entry NP_060568.3 (GeneID 1894).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 1894
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.