missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ECSIT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560183
This item is not returnable.
View return policy
Description
Immunogen sequence: HMEPDLSARV TIYQVPLPKD STGAADPPQP HIVGIQSPDQ QAALARHNPA RPVFVEGPFS LWLRNKCVYY HILRADLLPP EEREVEETPE EWNLYYPM Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 85%.
Activation of NF-kappa-B as a result of Toll-like receptor (TLR) and IL-1 receptor signaling is a major component of innate immune responses. Signals from these receptors are relayed by a number of adapter molecules such as TRIF, TIRAP, and MyD88 to kinases such as IRAK and other intermediates such as TNF receptor associated factor (TRAF)-6. ECSIT (evolutionarily conserved signaling intermediate in Toll pathways) was initially identified as a cytoplasmic protein interacting specifically with TNF receptor associated factor (TRAF)-6 in the TLR pathway. Recently however, ECSIT has also been shown to be required for bone morphogenetic protein (Bmp) signaling and mesoderm formation during mouse embryogenesis, indicating the possibility of cross-talk between the TLR/IL-B and Bmp signaling pathways.
Specifications
| ECSIT | |
| Polyclonal | |
| Unconjugated | |
| ECSIT | |
| Ecsit; ECSIT homolog; ECSIT homolog (Drosophila); ECSIT signalling integrator; evolutionarily conserved signaling intermediate in Toll pathway, mitochondrial; likely ortholog of mouse signaling intermediate in Toll pathway evolutionarily conserved; Protein SITPEC; signaling intermediate in Toll pathway-evolutionarily conserved; Sitpec | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26940, 300447, 51295 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5XIC2, Q9BQ95, Q9QZH6 | |
| ECSIT | |
| Recombinant protein corresponding to Human ECSIT. Recombinant protein control fragment (Product #RP-99074). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction