missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ECRG4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572717
This item is not returnable.
View return policy
Description
ECRG4 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
Probable hormone that may attenuate cell proliferation and induce senescence of oligodendrocyte and neural precursor cells in the central nervous system. ECRG4-induced senescence is characterized by G1 arrest, RB1 dephosphorylation and accelerated CCND1 and CCND3 proteasomal degradation. [UniProt].
Specifications
| ECRG4 | |
| Polyclonal | |
| Unconjugated | |
| C2orf40 | |
| Augurin; C2orf40; chromosome 2 open reading frame 40; ECRG4; esophageal cancer related gene 4 protein; esophageal cancer-related gene 4 protein; UNQ761/PRO1508 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84417 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q9H1Z8 | |
| C2orf40 | |
| ECRG4 antibody for a 20 amino acid synthetic peptide near the carboxy terminus of human ECRG4. The immunogen is within the last 50 amino acids of ECRG4. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction