missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ECRG4 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA552576

Product Code. 15700531

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: KREAPVPTKT KVAVDENKAK EFLGSLKRQK RQLWDRTRPE VQQWYQQFLY MGFDEAKFED DITYWLNRDR NGHEYYGDYY QRHYDEDSAI GPRSPYGFRH GASVNYD Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 85%.

Probable hormone that may attenuate cell proliferation and induce senescence of oligodendrocyte and neural precursor cells in the central nervous system. ECRG4-induced senescence is characterized by G1 arrest, RB1 dephosphorylation and accelerated CCND1 and CCND3 proteasomal degradation. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ECRG4
Polyclonal
Unconjugated
C2orf40
Augurin; C2orf40; chromosome 2 open reading frame 40; ECRG4; esophageal cancer related gene 4 protein; esophageal cancer-related gene 4 protein; UNQ761/PRO1508
Rabbit
Antigen Affinity Chromatography
RUO
84417
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9H1Z8
C2orf40
Recombinant protein corresponding to Human ECRG4. Recombinant protein control fragment (Product #RP-90147).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.