missing translation for 'onlineSavingsMsg'
Learn More

ECRG4 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18413198 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18413198 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18413198 Supplier Novus Biologicals Supplier No. NBP281925AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ECRG4 Polyclonal antibody specifically detects ECRG4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ECRG4
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias chromosome 2 open reading frame 40, ECRG4augurin, esophageal cancer related gene 4 protein, Esophageal cancer-related gene 4 protein
Host Species Rabbit
Immunogen ECRG4 antibody was raised against a 20 amino acid synthetic peptide near the carboxy terminus of human ECRG4. The immunogen is located within the last 50 amino acids of ECRG4. Amino Acid Squence: DRNHEYTGDYYQRHYDEDS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 84417
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.