missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ecopladib, MedChemExpress
Description
Ecopladib is a sub-micromolar inhibitor of cytosolic phospholipase A2α (cPLA2α), with IC50s of 0.15 μM and 0.11 μM in the GLU micelle and rat whole blood assays, respectively.
Specifications
Specifications
| Chemical Name or Material | Ecopladib |
| CAS | 381683-92-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C39H33Cl3N2O5S |
| Quantity | 1 mg |
| Synonym | PLA 725 |
| SMILES | O=C(O)C1=CC=C(OCCC2=C(CCNS(=O)(CC3=CC=C(Cl)C(Cl)=C3)=O)N(C(C4=CC=CC=C4)C5=CC=CC=C5)C6=C2C=C(Cl)C=C6)C=C1 |
| Molecular Weight (g/mol) | 748.11 |
| Formula Weight | 748.11 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?