Learn More
Cole Parmer™ Economical Polypropylene Graduated Cylinder
Polypropylene (PP) cylinders offer very good chemical resistance.
£15.21 - £22.36
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
15777968
|
Cole Parmer™
06137-70 |
£15.21
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||||
15787968
|
Cole Parmer™
06137-72 |
25 mL |
£15.21
Each |
In Stock
Log in to see stock. |
|||||
15797968
|
Cole Parmer™
06137-74 |
£19.68
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||||
15707978
|
Cole Parmer™
06137-76 |
£22.36
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||||||
Description
- Meet ISO Standard 6706.
- Calibrated to contain for measuring final volumes.
- Molded graduations for durability.
- Polypropylene construction provides very good chemical resistance.
Specifications
Polypropylene |