Learn More
Cole Parmer™ Economical Polypropylene Graduated Cylinder
Polypropylene (PP) cylinders offer very good chemical resistance.
Brand: Cole Parmer™ 06137-70
Code : NEW
Additional Details : Weight : 0.20000kg
Description
- Meet ISO Standard 6706.
- Calibrated to contain for measuring final volumes.
- Molded graduations for durability.
- Polypropylene construction provides very good chemical resistance.
Specifications
10 mL | |
ISO 6706 | |
Autoclavable | |
Hexagonal |
Polypropylene | |
To Contain | |
Round | |
Graduated Cylinder |