missing translation for 'onlineSavingsMsg'
Learn More

Econazole, TRC

Product Code. 30371195
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 30371195

Brand: Toronto Research Chemicals TRCE326003250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Econazole
CAS 27220-47-9
Molecular Formula C18 H15 Cl3 N2 O
Quantity 250 mg
Synonym Econazole, 1-[(2RS)-2-[(4-Chlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole, 1H-Imidazole, 1-[2-[(4-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]-, Imidazole, 1-[2,4-dichloro-β-[(p-chlorobenzyl)oxy]phenethyl]- (8CI), (±)-Econazole, 1-[2,4-Dichloro-β-[(p-chlorobenzyl)oxy]phenethyl]imidazole, Econazole, NSC 187789
InChI Formula InChI=1S/C18H15Cl3N2O/c19-14-3-1-13(2-4-14)11-24-18(10-23-8-7-22-12-23)16-6-5-15(20)9-17(16)21/h1-9,12,18H,10-11H2
SMILES Clc1ccc(COC(Cn2ccnc2)c3ccc(Cl)cc3Cl)cc1
IUPAC Name 1-[2-[(4-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]imidazole
Molecular Weight (g/mol) 381.68
Formula Weight 380.025
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado